Assay Details
| Gene Symbol: brca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110394.2 | NP_001103864.2 | 26 - 27 | 8473 | 72 | |
| XM_005157363.3 | XP_005157420.1 | 26 - 27 | 8610 | 72 | ||
| GenBank mRNA | AL730371.1 | - | 2 - 3 | 390 | 72 | |
| BC171710.1 | - | 26 - 27 | 8351 | 72 | ||
| EF088196.1 | - | 26 - 27 | 8470 | 72 | ||
More Information
Set Membership: |
|

brca2
Genomic Map