Assay Details
| Gene Symbol: isl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130962.1 | NP_571037.1 | 3 - 4 | 684 | 57 | |
| XM_021475997.1 | XP_021331672.1 | 3 - 4 | 725 | 57 | ||
| GenBank mRNA | BC060892.1 | - | 3 - 4 | 648 | 57 | |
| D21135.1 | - | 3 - 4 | 684 | 57 | ||
More Information
Set Membership: |
|

isl1
Genomic Map