Assay Details
Gene Symbol: tbxta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131162.1 | NP_571237.1 | 7 - 8 | 1087 | 61 | |
GenBank mRNA | BC162297.1 | - | 7 - 8 | 1082 | 61 | |
BC162300.1 | - | 7 - 8 | 1082 | 61 | ||
S57147.1 | - | 7 - 8 | 1087 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
