Assay Details
| Gene Symbol: pla2g4aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131295.2 | NP_571370.1 | 11 - 12 | 1269 | 127 | |
| GenBank mRNA | BC162082.1 | - | 10 - 11 | 1197 | 127 | |
| U10330.1 | - | 11 - 12 | 1270 | 127 | ||
More Information
Set Membership: |
|

pla2g4aa
Genomic Map