Assay Details
| Gene Symbol: pla2g4aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131295.2 | NP_571370.1 | 17 - 18 | 2196 | 95 | |
| GenBank mRNA | BC162082.1 | - | 16 - 17 | 2124 | 95 | |
| U10330.1 | - | 17 - 18 | 2197 | 95 | ||
More Information
Set Membership: |
|

pla2g4aa
Genomic Map