Assay Details
| Gene Symbol: notch1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131302.2 | NP_571377.2 | 16 - 17 | 2670 | 89 | |
| GenBank mRNA | AY805407.1 | - | 16 - 17 | 2670 | 89 | |
More Information
Set Membership: |
|

notch1b
Genomic Map