Assay Details
Gene Symbol: notch1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131302.2 | NP_571377.2 | 2 - 3 | 272 | 67 | |
XM_005168649.4 | XP_005168706.1 | 2 - 3 | 369 | 67 | ||
GenBank mRNA | AY805407.1 | - | 2 - 3 | 272 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
