Assay Details
Gene Symbol: notch1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131302.2 | NP_571377.2 | 4 - 5 | 874 | 110 | |
GenBank mRNA | AY805407.1 | - | 4 - 5 | 874 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
