Assay Details
| Gene Symbol: notch1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131302.2 | NP_571377.2 | 7 - 8 | 1298 | 77 | |
| XM_005168649.4 | XP_005168706.1 | 6 - 7 | 1326 | 77 | ||
| GenBank mRNA | AY805407.1 | - | 7 - 8 | 1298 | 77 | |
More Information
Set Membership: |
|

notch1b
Genomic Map