Assay Details
Gene Symbol: hspe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328390.1 | NP_001315319.1 | 4 - 5 | 561 | 74 | |
NM_001328391.1 | NP_001315320.1 | 3 - 4 | 429 | 74 | ||
NM_131526.2 | NP_571601.1 | 4 - 5 | 485 | 74 | ||
GenBank mRNA | BC071419.1 | - | 3 - 4 | 339 | 74 | |
EH434761.1 | - | 3 - 4 | 398 | 74 | ||
EH445965.1 | - | 3 - 4 | 335 | 74 | ||
EH546702.1 | - | 4 - 5 | 516 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
