Assay Details
| Gene Symbol: rpl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173235.3 | NP_775342.2 | 5 - 6 | 518 | 93 | |
| GenBank mRNA | AY099532.1 | - | 5 - 6 | 420 | 93 | |
| BC059530.1 | - | 4 - 5 | 401 | 93 | ||
| EH450194.1 | - | 5 - 6 | 448 | 93 | ||
| EH544688.1 | - | 5 - 6 | 557 | 93 | ||
More Information
Set Membership: |
|

rpl24
Genomic Map