Assay Details
| Gene Symbol: opn4.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178289.3 | NP_840074.2 | 1 | 1539 | 69 | |
| GenBank mRNA | AY078161.1 | - | 1 | 69 | ||
| BC116614.1 | - | 1 | 1539 | 69 | ||
| GQ925716.1 | - | 1 | 1422 | 69 | ||
| KT008429.1 | - | 1 | 1422 | 69 | ||
More Information
Set Membership: |
|

opn4.1
Genomic Map