Assay Details
| Gene Symbol: anxa11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181765.2 | NP_861430.2 | 10 - 11 | 1228 | 83 | |
| NM_183410.2 | NP_899670.1 | 10 - 11 | 1099 | 83 | ||
| GenBank mRNA | AY178801.1 | - | 10 - 11 | 1220 | 83 | |
| BC053208.1 | - | 10 - 11 | 1061 | 83 | ||
More Information
Set Membership: |
|

anxa11a
Genomic Map