Assay Details
| Gene Symbol: anxa11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181765.2 | NP_861430.2 | 11 - 12 | 1320 | 58 | |
| NM_183410.2 | NP_899670.1 | 11 - 12 | 1191 | 58 | ||
| GenBank mRNA | AY178801.1 | - | 11 - 12 | 1312 | 58 | |
| BC053208.1 | - | 11 - 12 | 1153 | 58 | ||
More Information
Set Membership: |
|

anxa11a
Genomic Map