Assay Details
Gene Symbol: f9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182868.2 | NP_878288.2 | 3 - 4 | 342 | 68 | |
GenBank mRNA | AF515270.1 | - | 3 - 4 | 270 | 68 | |
EH506608.1 | - | 3 - 4 | 285 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
