Assay Details
Gene Symbol: f9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182868.2 | NP_878288.2 | 4 - 5 | 457 | 92 | |
GenBank mRNA | AF515270.1 | - | 4 - 5 | 385 | 92 | |
EH506608.1 | - | 4 - 5 | 400 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
