Assay Details
Gene Symbol: f9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182868.2 | NP_878288.2 | 6 - 7 | 898 | 95 | |
XM_005173201.4 | XP_005173258.1 | 4 - 5 | 631 | 95 | ||
GenBank mRNA | AF515270.1 | - | 6 - 7 | 826 | 95 | |
EH532764.1 | - | 2 - 3 | 820 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
