Assay Details
Gene Symbol: tgfb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182873.1 | NP_878293.1 | 3 - 4 | 842 | 66 | |
GenBank mRNA | AY178450.1 | - | 3 - 4 | 842 | 66 | |
BC162361.1 | - | 3 - 4 | 751 | 66 | ||
BC162366.1 | - | 3 - 4 | 751 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
