Assay Details
| Gene Symbol: slc4a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198338.1 | NP_938152.1 | 10 - 11 | 937 | 66 | |
| GenBank mRNA | AF350072.1 | - | 10 - 11 | 937 | 66 | |
| BC093126.1 | - | 3 - 4 | 238 | 66 | ||
| BC163932.1 | - | 10 - 11 | 923 | 66 | ||
More Information
Set Membership: |
|

slc4a1a
Genomic Map