Assay Details
| Gene Symbol: slc4a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198338.1 | NP_938152.1 | 11 - 12 | 1146 | 75 | |
| GenBank mRNA | AF350072.1 | - | 11 - 12 | 1146 | 75 | |
| AY098936.1 | - | 11 - 12 | 1179 | 75 | ||
| BC093126.1 | - | 4 - 5 | 447 | 75 | ||
| BC163932.1 | - | 11 - 12 | 1132 | 75 | ||
More Information
Set Membership: |
|

slc4a1a
Genomic Map