Assay Details
| Gene Symbol: slc4a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198338.1 | NP_938152.1 | 17 - 18 | 2196 | 103 | |
| GenBank mRNA | AF350072.1 | - | 17 - 18 | 2196 | 103 | |
| AY098936.1 | - | 17 - 18 | 2229 | 103 | ||
| BC124121.1 | - | 3 - 4 | 204 | 103 | ||
| BC134843.1 | - | 4 - 5 | 529 | 103 | ||
| BC163932.1 | - | 17 - 18 | 2182 | 103 | ||
More Information
Set Membership: |
|

slc4a1a
Genomic Map