Assay Details
| Gene Symbol: slc4a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198338.1 | NP_938152.1 | 18 - 19 | 2436 | 101 | |
| GenBank mRNA | AF350072.1 | - | 18 - 19 | 2436 | 101 | |
| BC124121.1 | - | 4 - 5 | 444 | 101 | ||
More Information
Set Membership: |
|

slc4a1a
Genomic Map