Assay Details
| Gene Symbol: ckmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198801.1 | NP_942096.1 | 5 - 6 | 759 | 105 | |
| XM_021470450.1 | XP_021326125.1 | 4 - 5 | 1047 | 105 | ||
| GenBank mRNA | BC048050.1 | - | 5 - 6 | 759 | 105 | |
| BC165565.1 | - | 4 - 5 | 696 | 105 | ||
More Information
Set Membership: |
|

ckmt1
Genomic Map