Assay Details
Gene Symbol: ckmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198801.1 | NP_942096.1 | 8 - 9 | 1113 | 72 | |
XM_021470450.1 | XP_021326125.1 | 7 - 8 | 1401 | 72 | ||
GenBank mRNA | BC048050.1 | - | 8 - 9 | 1113 | 72 | |
BC165565.1 | - | 7 - 8 | 1050 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
