Assay Details
| Gene Symbol: zhx2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198814.2 | NP_942109.2 | 3 - 4 | 2802 | 69 | |
| GenBank mRNA | BC141782.1 | - | 3 - 4 | 2802 | 69 | |
More Information
Set Membership: |
|

zhx2a
Genomic Map