Assay Details
Gene Symbol: kcnd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199802.1 | NP_956096.1 | 6 - 7 | 2074 | 65 | |
XM_005167002.4 | XP_005167059.1 | 7 - 8 | 2310 | 65 | ||
XM_005167003.4 | XP_005167060.1 | 7 - 8 | 2307 | 65 | ||
GenBank mRNA | BC045304.1 | - | 6 - 7 | 2074 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
