Assay Details
| Gene Symbol: mlst8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320388.1 | NP_001307317.1 | 8 - 9 | 1003 | 65 | |
| GenBank mRNA | BC044176.1 | - | 8 - 9 | 998 | 65 | |
| BC068352.1 | - | 8 - 9 | 1003 | 65 | ||
| BC164355.1 | - | 7 - 8 | 895 | 65 | ||
| EH568073.1 | - | 1 - 2 | 360 | 65 | ||
More Information
Set Membership: |
|

mlst8
Genomic Map