Assay Details
| Gene Symbol: seh1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199923.2 | NP_956217.2 | 2 - 3 | 389 | 56 | |
| XM_021468000.1 | XP_021323675.1 | 2 - 3 | 384 | 56 | ||
| GenBank mRNA | AY398414.1 | - | 2 - 3 | 389 | 56 | |
| BC051159.1 | - | 2 - 3 | 319 | 56 | ||
| CT658356.2 | - | 2 - 3 | 263 | 56 | ||
More Information
Set Membership: |
|

seh1l
Genomic Map