Assay Details
Gene Symbol: pip4k2ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200101.2 | NP_956395.2 | 4 - 5 | 503 | 80 | |
GenBank mRNA | BC045883.1 | - | 4 - 5 | 640 | 80 | |
BC071466.1 | - | 4 - 5 | 503 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
