Assay Details
Gene Symbol: crispld1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200470.1 | NP_956764.1 | 6 - 7 | 961 | 100 | |
XM_009298567.3 | XP_009296842.1 | 6 - 7 | 984 | 100 | ||
GenBank mRNA | BC055176.1 | - | 6 - 7 | 961 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
