Assay Details
| Gene Symbol: naa15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200646.2 | NP_956940.2 | 16 - 17 | 2262 | 120 | |
| GenBank mRNA | BC045491.1 | - | 16 - 17 | 2231 | 120 | |
| BC057466.1 | - | 16 - 17 | 2262 | 120 | ||
| BC165243.1 | - | 16 - 17 | 2095 | 120 | ||
More Information
Set Membership: |
|

naa15a
Genomic Map