Assay Details
| Gene Symbol: lsm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200696.1 | NP_956990.1 | 2 - 3 | 162 | 62 | |
| GenBank mRNA | BC059435.1 | - | 2 - 3 | 162 | 62 | |
| BC062285.1 | - | 2 - 3 | 143 | 62 | ||
More Information
Set Membership: |
|

lsm4
Genomic Map