Assay Details
| Gene Symbol: ndufa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200968.1 | NP_957262.1 | 2 - 3 | 261 | 89 | |
| GenBank mRNA | BC042332.1 | - | 2 - 3 | 261 | 89 | |
More Information
Set Membership: |
|

ndufa6
Genomic Map