Assay Details
| Gene Symbol: ephx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201068.1 | NP_957362.1 | 8 - 9 | 1224 | 57 | |
| GenBank mRNA | BC045930.1 | - | 8 - 9 | 1224 | 57 | |
| BC165341.1 | - | 7 - 8 | 1195 | 57 | ||
More Information
Set Membership: |
|

ephx1
Genomic Map