Assay Details
| Gene Symbol: capza1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212697.1 | NP_997862.1 | 4 - 5 | 248 | 96 | |
| GenBank mRNA | BC047860.1 | - | 4 - 5 | 248 | 96 | |
| BC067561.1 | - | 4 - 5 | 228 | 96 | ||
More Information
Set Membership: |
|

capza1a
Genomic Map