Assay Details
Gene Symbol: pgp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212726.2 | NP_997891.2 | 1 - 2 | 810 | 66 | |
GenBank mRNA | AY391472.1 | - | 1 - 2 | 700 | 66 | |
BC045860.1 | - | 1 - 2 | 758 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
