Assay Details
| Gene Symbol: mettl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212780.1 | NP_997945.1 | 4 - 5 | 1018 | 57 | |
| GenBank mRNA | BC067182.1 | - | 4 - 5 | 1018 | 57 | |
More Information
Set Membership: |
|

mettl3
Genomic Map