Assay Details
| Gene Symbol: kirrel1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001362419.1 | NP_001349348.1 | 12 - 13 | 1772 | 139 | |
| NM_212868.2 | NP_998033.1 | 12 - 13 | 1772 | 139 | ||
| GenBank mRNA | BC066766.1 | - | 12 - 13 | 1772 | 139 | |
More Information
Set Membership: |
|

kirrel1a
Genomic Map