Assay Details
| Gene Symbol: kirrel1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212868.2 | NP_998033.1 | 13 - 14 | 1891 | 62 | |
| GenBank mRNA | BC066766.1 | - | 13 - 14 | 1891 | 62 | |
More Information
Set Membership: |
|

kirrel1a
Genomic Map