Assay Details
| Gene Symbol: znf207a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213197.1 | NP_998362.1 | 2 - 3 | 222 | 63 | |
| XM_005164009.4 | XP_005164066.1 | 2 - 3 | 258 | 63 | ||
| XM_009299526.3 | XP_009297801.1 | 2 - 3 | 258 | 63 | ||
| GenBank mRNA | BC065456.1 | - | 2 - 3 | 222 | 63 | |
More Information
Set Membership: |
|

znf207a
Genomic Map