Assay Details
Gene Symbol: ccni | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213221.1 | NP_998386.1 | 5 - 6 | 702 | 60 | |
GenBank mRNA | BC044400.1 | - | 5 - 6 | 690 | 60 | |
BC068369.1 | - | 5 - 6 | 702 | 60 | ||
BC129229.1 | - | 4 - 5 | 471 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
