Assay Details
| Gene Symbol: rp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213446.1 | NP_998611.1 | 2 - 3 | 1009 | 78 | |
| XM_005165977.1 | XP_005166034.1 | 2 - 3 | 1035 | 78 | ||
| XM_005165978.1 | XP_005166035.1 | 2 - 3 | 1035 | 78 | ||
| GenBank mRNA | BC046879.1 | - | 2 - 3 | 1009 | 78 | |
| BC164845.1 | - | 1 - 2 | 683 | 78 | ||
| HQ641392.1 | - | 2 - 3 | 881 | 78 | ||
More Information
Set Membership: |
|

rp2
Genomic Map