Assay Details
| Gene Symbol: rp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213446.1 | NP_998611.1 | 5 - 6 | 1315 | 90 | |
| XM_005165978.1 | XP_005166035.1 | 5 - 6 | 1341 | 90 | ||
| GenBank mRNA | BC046879.1 | - | 5 - 6 | 1315 | 90 | |
More Information
Set Membership: |
|

rp2
Genomic Map