Assay Details
Gene Symbol: fbxw11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213504.2 | NP_998669.2 | 11 - 12 | 1462 | 68 | |
XM_005155294.4 | XP_005155351.1 | 11 - 12 | 1473 | 68 | ||
GenBank mRNA | BC056809.1 | - | 11 - 12 | 1464 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
