Assay Details
Gene Symbol: gpm6ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214687.1 | NP_999852.1 | 4 - 5 | 552 | 65 | |
XM_005159981.4 | XP_005160038.1 | 4 - 5 | 1045 | 65 | ||
XM_005159982.3 | XP_005160039.1 | 4 - 5 | 628 | 65 | ||
GenBank mRNA | AY070264.1 | - | 4 - 5 | 552 | 65 | |
AY070266.1 | - | 4 - 5 | 552 | 65 | ||
BC075774.1 | - | 4 - 5 | 636 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
