Assay Details
| Gene Symbol: adcyap1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272081.1 | NP_001259010.1 | 2 - 3 | 445 | 75 | |
| NM_214715.2 | NP_999880.1 | 2 - 3 | 445 | 75 | ||
| GenBank mRNA | AF329730.1 | - | 2 - 3 | 388 | 75 | |
| BC083192.1 | - | 2 - 3 | 413 | 75 | ||
| BC083516.1 | - | 2 - 3 | 362 | 75 | ||
| BC164866.1 | - | 1 - 2 | 139 | 75 | ||
| EH439402.1 | - | 2 - 3 | 395 | 75 | ||
| EH442184.1 | - | 2 - 3 | 430 | 75 | ||
More Information
Set Membership: |
|

adcyap1b
Genomic Map