Assay Details
Gene Symbol: dhdh.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214735.1 | NP_999900.1 | 3 - 4 | 519 | 132 | |
GenBank mRNA | BC053221.1 | - | 3 - 4 | 519 | 132 | |
BC164134.1 | - | 3 - 4 | 403 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
