Assay Details
| Gene Symbol: dhdh.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214735.1 | NP_999900.1 | 6 - 7 | 1050 | 79 | |
| GenBank mRNA | BC053221.1 | - | 6 - 7 | 1050 | 79 | |
| BC164134.1 | - | 6 - 7 | 934 | 79 | ||
More Information
Set Membership: |
|

dhdh.1
Genomic Map