Assay Details
Gene Symbol: puf60b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002121.1 | NP_001002121.1 | 3 - 4 | 194 | 98 | |
XM_009303395.3 | XP_009301670.1 | 3 - 4 | 332 | 98 | ||
GenBank mRNA | BC071467.1 | - | 3 - 4 | 194 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
