Assay Details
Gene Symbol: med22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002144.1 | NP_001002144.1 | 3 - 4 | 264 | 84 | |
XM_005171831.2 | XP_005171888.1 | 3 - 4 | 331 | 84 | ||
XM_005171832.4 | XP_005171889.1 | 3 - 4 | 441 | 84 | ||
XM_021476063.1 | XP_021331738.1 | 3 - 4 | 441 | 84 | ||
GenBank mRNA | BC049480.1 | - | 3 - 4 | 508 | 84 | |
BC071397.1 | - | 3 - 4 | 264 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
